4-Bromo-8-chloroquinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2311-1G HC-2311-5G
COBBHC-2311-1GEA 0 GBP
COBBHC-2311-1G COBBHC-2311-5G
4-Bromo-8-chloroquinoline
4-Bromo-8-chloroquinoline
Formula: C₉H₅BrClN
MDL Number: MFCD07700233
CAS Number: 927800-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR