3-Quinolineboronic acid hydrochloride

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-2394-5G BB-2394-1G
COBBBB-2394-5GEA 0 GBP
COBBBB-2394-5G COBBBB-2394-1G
3-Quinolineboronic acid hydrochloride
3-Quinolineboronic acid hydrochloride
Formula: C₉H₉BClNO₂
MDL Number: MFCD06659836
CAS Number: 850568-71-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR