6-Quinolineboronic acid pinacol ester, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Quinoline-6-boronic acid Pinacol ester, 6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline, Quinoline-6-boronic acid, pinacol ester, 2-(Quinolin-6-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

M-Clarity™ quality level = MQ100

641618-1G 641618-5G
SIAL641618-1GEA 77.5 GBP
SIAL641618-1G SIAL641618-5G
6-Quinolineboronic acid pinacol ester, Sigma-Aldrich®
6-Quinolineboronic acid pinacol ester
Formula: C₁₅H₁₈BNO₂
MDL Number: MFCD06411327
CAS Number: 406463-06-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR