3-Amino-N-benzylbenzenesulphonamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-0127-5G OT-0127-1G
COBBOT-0127-5GEA 0 GBP
COBBOT-0127-5G COBBOT-0127-1G
3-Amino-N-benzylbenzenesulphonamide
3-Amino-N-benzylbenzenesulphonamide
Formula: C₁₃H₁₄N₂O₂S
MDL Number: MFCD07363817
CAS Number: 303780-52-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR