N-Benzyl-4-fluorobenzenesulphonamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-1116-5G OT-1116-1G
COBBOT-1116-5GEA 0 GBP
COBBOT-1116-5G COBBOT-1116-1G
N-Benzyl-4-fluorobenzenesulphonamide
N-Benzyl 4-fluorobenzenesulphonamide
Formula: C₁₃H₁₂FNO₂S
MDL Number: MFCD01213083
CAS Number: 727-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR