2-Benzyloxy-5-fluorophenylboronic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

016102-1G 016102-5G
FLUO016102-1GEA 0 GBP
FLUO016102-1G FLUO016102-5G
2-Benzyloxy-5-fluorophenylboronic acid
2-Benzyloxy-5-fluorophenylboronic acid
Formula: C₁₃H₁₂BFO₃
MDL Number: MFCD03788403
CAS Number: 779331-47-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR