4-Benzyloxy-3-fluorophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Benzyloxy-3-fluorobenzeneboronic acid

BB-2688-5G BB-2688-25G BB-2688-1G BB-2688-100G
COBBBB-2688-5GEA 0 GBP
COBBBB-2688-5G COBBBB-2688-25G COBBBB-2688-1G COBBBB-2688-100G
4-Benzyloxy-3-fluorophenylboronic acid
4-Benzyloxy-3-fluorophenylboronic acid
Formula: C₁₃H₁₂BFO₃
MW: 246.04 g/mol
MDL Number: MFCD00994627
CAS Number: 133057-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR