5-Benzyloxy-2-fluorophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-(Benzyloxy)-2-fluorophenylboronic acid

BB-5631-1G BB-5631-5G BB-5631-250M
COBBBB-5631-1GEA 0 GBP
COBBBB-5631-1G COBBBB-5631-5G COBBBB-5631-250M
5-Benzyloxy-2-fluorophenylboronic acid
5-Benzyloxy-2-fluorophenylboronic acid
Formula: C₁₃H₁₂BFO₃
MDL Number: MFCD13195645
CAS Number: 1217500-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR