5-Carbamoyl-2-fluorophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (5-Carbamoyl-2-fluorophenyl)boronic acid, 5-Carbamoyl-2-fluorophenyboronic acid, 5-Carbamoyl-2-fluorobenzeneboronic acid

BB-3334-5G BB-3334-1G
COBBBB-3334-5GEA 0 GBP
COBBBB-3334-5G COBBBB-3334-1G
5-Carbamoyl-2-fluorophenylboronic acid
5-Carbamoyl-2-fluorophenylboronic acid
Formula: C₇H₇BFNO₃
MW: 182.95 g/mol
MDL Number: MFCD08436055
CAS Number: 874289-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR