3-Carbamoyl-5-chlorophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (3-Carbamoyl-5-chlorophenyl)boronic acid

BB-3238-5G BB-3238-1G
COBBBB-3238-5GEA 0 GBP
COBBBB-3238-5G COBBBB-3238-1G
3-Carbamoyl-5-chlorophenylboronic acid
3-Carbamoyl-5-chlorophenylboronic acid
Formula: C₇H₇BClNO₃
MW: 199.4000 g/mol
MDL Number: MFCD09800858
CAS Number: 957120-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR