Methyl-3-(benzyloxy)-4-bromobenzoate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4138-5G CA-4138-1G
COBBCA-4138-5GEA 0 GBP
COBBCA-4138-5G COBBCA-4138-1G
Methyl-3-(benzyloxy)-4-bromobenzoate
Methyl 3-(benzyloxy)-4-bromobenzoate
Formula: C₁₅H₁₃BrO₃
MDL Number: MFCD08436016
CAS Number: 17054-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR