N-Benzyl-2-bromo-N-methylbenzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CS-9419-5G CS-9419-1G
COBBCS-9419-5GEA 0 GBP
COBBCS-9419-5G COBBCS-9419-1G
N-Benzyl-2-bromo-N-methylbenzamide
N-Benzyl-2-bromo-N-methylbenzamide
Formula: C₁₅H₁₄BrNO
MDL Number: MFCD00784541
CAS Number: 124461-28-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR