5-Bromo-2-chloro-N-(4-methoxybenzyl)benzamide
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4725-1G
CA-4725-5G
CA-4725-25G
COBBCA-4725-1GEA
0
GBP
COBBCA-4725-1G
COBBCA-4725-5G
COBBCA-4725-25G
5-Bromo-2-chloro-N-(4-methoxybenzyl)benzamide
5-Bromo-2-chloro-N-(4-methoxybenzyl)benzamide
Formula:
C₁₅H₁₃BrClNO₂ |
MDL Number:
MFCD04219846 CAS Number: 701255-27-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...