3-(Benzyloxy)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (3-(Benzyloxy)phenyl)boronic acid, 3-Benzyloxybenzeneboronic acid

BB-2153-25G BB-2153-5G
COBBBB-2153-25GEA 0 GBP
COBBBB-2153-25G COBBBB-2153-5G
3-(Benzyloxy)phenylboronic acid
3-(Benzyloxy)phenylboronic acid
Formula: C₁₃H₁₃BO₃
MW: 228.06 g/mol
MDL Number: MFCD02093063
CAS Number: 156682-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR