2-(Benzyloxy)phenylboronic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Benzyloxyphenylboronic acid, (2-(Benzyloxy)phenyl)boronic acid, 2-Benzyloxybenzeneboronic acid

016104-1G 016104-25G 016104-5G
FLUO016104-1GEA 0 GBP
FLUO016104-1G FLUO016104-25G FLUO016104-5G
2-(Benzyloxy)phenylboronic acid
2-(Benzyloxy)phenylboronic acid
Formula: C₁₃H₁₃BO₃
MDL Number: MFCD01632206
CAS Number: 190661-29-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR