4-(Benzyloxy)phenylboronic acid pinacol ester

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Benzyloxybenzeneboronic acid pinacol ester, 2-(4-(Benzyloxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

AS21519G1 AS21519G5
ACTIAS21519G1EA 0 GBP
ACTIAS21519G1 ACTIAS21519G5
4-(Benzyloxy)phenylboronic acid pinacol ester
4-(Benzyloxy)phenylboronic acid pinacol ester
Formula: C₁₉H₂₃BO₃
MW: 310.19 g/mol
MDL Number: MFCD04974107
CAS Number: 754226-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR