2-(Benzyloxy)phenylboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PN-2466-5G PN-2466-1G
COBBPN-2466-5GEA 0 GBP
COBBPN-2466-5G COBBPN-2466-1G
2-(Benzyloxy)phenylboronic acid pinacol ester
2-(Benzyloxy)phenylboronic acid pinacol ester
Formula: C₁₉H₂₃BO₃
MDL Number: MFCD03001334
CAS Number: 1027757-13-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR