2-(4-Morpholino)-5-nitropyridine ≥95%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

H34452.03 H34452.06
H34452.03EA 40.5 GBP
H34452.03 H34452.06
2-(4-Morpholino)-5-nitropyridine ≥95%
2-(4-Morpholino)-5-nitropyridine

Purity: 95%
CAS No.: 26820-62-2
Molecular Formula: C9H11N3O3
Formula Weight: 209.20
Melting Point: 142-144°
Beilstein Reference: 0225480

Formula: C₉H₁₁N₃O₃
MDL Number: MFCD00023308
CAS Number: 26820-62-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR