N-Benzyl-3-bromo-5-methylbenzenesulphonamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6362-5G HC-6362-1G
COBBHC-6362-5GEA 0 GBP
COBBHC-6362-5G COBBHC-6362-1G
N-Benzyl-3-bromo-5-methylbenzenesulphonamide
N-Benzyl 3-bromo-5-methylbenzenesulphonamide
Formula: C₁₄H₁₄BrNO₂S
MDL Number: MFCD09972139
CAS Number: 1020252-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR