Methyl-3-iodo-4-methylbenzoate
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Methyl 3-iodo-4-methylbenzoate
AS23639G100
AS23639G25
ACTIAS23639G100EA
124
GBP
ACTIAS23639G100
ACTIAS23639G25
Methyl-3-iodo-4-methylbenzoate
Methyl 3-iodo-4-methylbenzoate
Formula:
C₉H₉IO₂ MW: 276.07 g/mol |
MDL Number:
MFCD00230583 CAS Number: 90347-66-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...