Methyl-3-iodo-2-methylbenzoate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 3-iodo-2-methylbenzoate

HC-3372-5G HC-3372-1G
COBBHC-3372-5GEA 0 GBP
COBBHC-3372-5G COBBHC-3372-1G
Methyl-3-iodo-2-methylbenzoate
Methyl 3-iodo-2-methylbenzoate
Formula: C₉H₉IO₂
MW: 276.0710 g/mol
MDL Number: MFCD12923250
CAS Number: 52570-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR