5-Chloro-3-methylanthranilic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Amino-5-chloro-3-methylbenzoic acid

M-Clarity™ quality level = MQ100

643513-25G 643513-5G
SIAL643513-25GEA 134 GBP
SIAL643513-25G SIAL643513-5G
5-Chloro-3-methylanthranilic acid, Sigma-Aldrich®
5-Chloro-3-methylanthranilic acid
Formula: C₈H₈ClNO₂
MW: 185.6076 g/mol
MDL Number: MFCD02358895
CAS Number: 20776-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR