5-Chloro-3-methylanthranilic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Amino-5-chloro-3-methylbenzoic acid

AS23247G100 AS23247G25
ACTIAS23247G100EA 0 GBP
ACTIAS23247G100 ACTIAS23247G25
5-Chloro-3-methylanthranilic acid
5-Chloro-3-methylanthranilic acid
Formula: C₈H₈ClNO₂
MW: 185.6076 g/mol
MDL Number: MFCD02358895
CAS Number: 20776-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR