3-Chloro-5-methyl-4-hydroxybenzonitrile
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS12574G1
AS12574G5
ACTIAS12574G1EA
0
GBP
ACTIAS12574G1
ACTIAS12574G5
3-Chloro-5-methyl-4-hydroxybenzonitrile
3-Chloro-5-methyl-4-hydroxybenzonitrile
Formula:
C₈H₆ClNO |
MDL Number:
MFCD08689583 CAS Number: 173900-45-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...