3-Bromo-N-cyclohexyl-4-methylbenzenesulphonamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-0161-5G OT-0161-1G
COBBOT-0161-5GEA 0 GBP
COBBOT-0161-5G COBBOT-0161-1G
3-Bromo-N-cyclohexyl-4-methylbenzenesulphonamide
3-Bromo-N-cyclohexyl-4-methylbenzenesulphonamide
Formula: C₁₃H₁₈BrNO₂S
MDL Number: MFCD07363834
CAS Number: 850429-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR