3-Bromo-N-butyl-5-methylbenzenesulphonamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6458-5G HC-6458-1G
COBBHC-6458-5GEA 0 GBP
COBBHC-6458-5G COBBHC-6458-1G
3-Bromo-N-butyl-5-methylbenzenesulphonamide
3-Bromo-N-butyl-5-methylbenzenesulphonamide
Formula: C₁₁H₁₆BrNO₂S
MDL Number: MFCD09972149
CAS Number: 1020252-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR