3-Bromo-N-4-dimethylbenzenesulphonamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-0173-5G OT-0173-1G
COBBOT-0173-5GEA 0 GBP
COBBOT-0173-5G COBBOT-0173-1G
3-Bromo-N-4-dimethylbenzenesulphonamide
3-Bromo-N,4-dimethylbenzenesulphonamide
Formula: C₈H₁₀BrNO₂S
MDL Number: MFCD09475907
CAS Number: 941294-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR