4-Bromo-N,N-3-trimethylbenzenesulphonamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-0530-5G OT-0530-1G
COBBOT-0530-5GEA 0 GBP
COBBOT-0530-5G COBBOT-0530-1G
4-Bromo-N,N-3-trimethylbenzenesulphonamide
4-Bromo-N,N,3-trimethylbenzenesulphonamide
Formula: C₉H₁₂BrNO₂S
MDL Number: MFCD09475820
CAS Number: 849532-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR