3-Bromo-N,N-dimethyl-5-methylbenzenesulphonamide
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-6456-5G
HC-6456-1G
COBBHC-6456-5GEA
0
GBP
COBBHC-6456-5G
COBBHC-6456-1G
3-Bromo-N,N-dimethyl-5-methylbenzenesulphonamide
3-Bromo-N,N-dimethyl-5-methylbenzenesulphonamide
Formula:
C₉H₁₂BrNO₂S |
MDL Number:
MFCD09972148 CAS Number: 1020252-92-9 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...