3-Bromo-N,N-diethyl-5-methylbenzenesulphonamide
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-6459-5G
HC-6459-1G
COBBHC-6459-5GEA
0
GBP
COBBHC-6459-5G
COBBHC-6459-1G
3-Bromo-N,N-diethyl-5-methylbenzenesulphonamide
3-Bromo-N,N-diethyl-5-methylbenzenesulphonamide
Formula:
C₁₁H₁₆BrNO₂S |
MDL Number:
MFCD09972150 CAS Number: 1020252-94-1 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...