4-Bromo-N-cyclopentyl-3-methylbenzamide
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4557-1G
CA-4557-5G
COBBCA-4557-1GEA
0
GBP
COBBCA-4557-1G
COBBCA-4557-5G
4-Bromo-N-cyclopentyl-3-methylbenzamide
4-Bromo-N-cyclopentyl-3-methylbenzamide
Formula:
C₁₃H₁₆BrNO |
MDL Number:
MFCD09972124 CAS Number: 1020252-78-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...