N-Methylbenzenesulphonamide ≥97%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

H55666.MD H55666.03
H55666.MDEA 33.2 GBP
H55666.MD H55666.03
N-Methylbenzenesulphonamide ≥97%
N-Methylbenzenesulphonamide

Purity: 97% CAS No.: 5183-78-8 Molecular Formula: C7H9NO2S Formula Weight: 171.21 MDL No.: MFCD00025023

Formula: C₇H₉NO₂S
MW: 171.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00025023
CAS Number: 5183-78-8
EINECS: 225-965-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR