N-Propyl 4-fluorobenzenesulphonamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-0957-1G OT-0957-5G
COBBOT-0957-1GEA 0 GBP
COBBOT-0957-1G COBBOT-0957-5G
N-Propyl 4-fluorobenzenesulphonamide
N-Propyl 4-fluorobenzenesulphonamide
Formula: C₉H₁₂FNO₂S
MDL Number: MFCD01212870
CAS Number: 433-05-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR