N-tert-Butyl-4-fluorobenzenesulphonamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-1118-5G OT-1118-1G
COBBOT-1118-5GEA 0 GBP
COBBOT-1118-5G COBBOT-1118-1G
N-tert-Butyl-4-fluorobenzenesulphonamide
N-tert-Butyl 4-fluorobenzenesulphonamide
Formula: C₁₀H₁₄FNO₂S
MDL Number: MFCD01213076
CAS Number: 29083-05-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR