4-Fluoro-2-methoxy-N-methylaniline hydrochloride

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-3046-5G HC-3046-1G
COBBHC-3046-5GEA 0 GBP
COBBHC-3046-5G COBBHC-3046-1G
4-Fluoro-2-methoxy-N-methylaniline hydrochloride
4-Fluoro-2-methoxy-N-methylaniline hydrochloride
Formula: C₈H₁₁ClFNO
MDL Number: MFCD13195693
CAS Number: 1215205-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR