N-Ethyl-4-fluorobenzamide
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4748-25G
CA-4748-1G
CA-4748-5G
COBBCA-4748-25GEA
0
GBP
COBBCA-4748-25G
COBBCA-4748-1G
COBBCA-4748-5G
N-Ethyl-4-fluorobenzamide
N-Ethyl 4-fluorobenzamide
Formula:
C₉H₁₀FNO |
MDL Number:
MFCD01214153 CAS Number: 772-18-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...