2-Cyano-4-fluorophenylboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(2-Cyano-4-fluorophenyl)boronic acid
BB-8788-1G
BB-8788-250M
COBBBB-8788-1GEA
0
GBP
COBBBB-8788-1G
COBBBB-8788-250M
2-Cyano-4-fluorophenylboronic acid
2-Cyano-4-fluorophenylboronic acid
Formula:
C₇H₅BFNO₂ MW: 164.9295 g/mol |
MDL Number:
MFCD12546526 CAS Number: 876601-43-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...