2-(Methylthio)-5-nitrobenzonitrile

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AN-1150-5G AN-1150-1G
COBBAN-1150-5GEA 0 GBP
COBBAN-1150-5G COBBAN-1150-1G
2-(Methylthio)-5-nitrobenzonitrile
2-(Methylthio)-5-nitrobenzonitrile
Formula: C₈H₆N₂O₂S
MDL Number: MFCD09475845
CAS Number: 74631-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR