4-(2-Aminoethoxy)benzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22873G1 AS22873G5
ACTIAS22873G1EA 141 GBP
ACTIAS22873G1 ACTIAS22873G5
4-(2-Aminoethoxy)benzonitrile
4-(2-Aminoethoxy)benzonitrile
Formula: C₉H₁₀N₂O
MDL Number: MFCD16495887
CAS Number: 67333-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR