2-(2-Aminoethoxy)benzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22875G1 AS22875G5
ACTIAS22875G1EA 168 GBP
ACTIAS22875G1 ACTIAS22875G5
2-(2-Aminoethoxy)benzonitrile
2-(2-Aminoethoxy)benzonitrile
Formula: C₉H₁₀N₂O
MDL Number: MFCD16495889
CAS Number: 57276-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR