2-Chlor-4-fluoro-5-(methoxycarbonyl)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-5106-1G BB-5106-500M BB-5106-250M
COBBBB-5106-1GEA 0 GBP
COBBBB-5106-1G COBBBB-5106-500M COBBBB-5106-250M
2-Chlor-4-fluoro-5-(methoxycarbonyl)phenylboronic acid
2-Chlor-4-fluoro-5-(methoxycarbonyl)phenylboronic acid
Formula: C₈H₇BClFO₄
MW: 232.401 g/mol
MDL Number: MFCD09878351
CAS Number: 957066-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR