4-Chloro-N,N-diisopropylbenzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4529-25G CA-4529-1G CA-4529-5G
COBBCA-4529-25GEA 0 GBP
COBBCA-4529-25G COBBCA-4529-1G COBBCA-4529-5G
4-Chloro-N,N-diisopropylbenzamide
4-Chloro-N,N-diisopropylbenzamide
Formula: C₁₃H₁₈ClNO
MW: 239.75 g/mol
MDL Number: MFCD00032340
CAS Number: 79606-45-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR