3-Chloro-5-(methylcarbamoyl)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-3236-1G BB-3236-5G
COBBBB-3236-1GEA 0 GBP
COBBBB-3236-1G COBBBB-3236-5G
3-Chloro-5-(methylcarbamoyl)phenylboronic acid
3-Chloro-5-(methylcarbamoyl)phenylboronic acid
Formula: C₈H₉BClNO₃
MDL Number: MFCD09800856
CAS Number: 957120-51-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR