3-Chloro-5-(dimethylcarbamoyl)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Chloro-5-(dimethylcarbonyl)phenylboronic acid, (3-Chloro-5-(dimethylcarbamoyl)phenyl)boronic acid

BB-3247-1G BB-3247-5G
COBBBB-3247-1GEA 0 GBP
COBBBB-3247-1G COBBBB-3247-5G
3-Chloro-5-(dimethylcarbamoyl)phenylboronic acid
3-Chloro-5-(dimethylcarbamoyl)phenylboronic acid
Formula: C₉H₁₁BClNO₃
MW: 227.4500 g/mol
MDL Number: MFCD09800861
CAS Number: 957120-57-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR