5-Bromo-N-cyclopropyl-2-methoxybenzenesulphonamide ≥97%
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OT-0304-5G
OT-0304-1G
COBBOT-0304-5GEA
0
GBP
COBBOT-0304-5G
COBBOT-0304-1G
5-Bromo-N-cyclopropyl-2-methoxybenzenesulphonamide ≥97%
5-Bromo-N-cyclopropyl-2-methoxybenzenesulphonamide
Formula:
C₁₀H₁₂BrNO₃S MW: 306.176 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08235093 CAS Number: 1000339-35-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...