5-Bromo-N-(2-hydroxyethyl)-2-methoxybenzenesulphonamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-0302-5G OT-0302-1G
COBBOT-0302-5GEA 0 GBP
COBBOT-0302-5G COBBOT-0302-1G
5-Bromo-N-(2-hydroxyethyl)-2-methoxybenzenesulphonamide
5-Bromo-N-(2-hydroxyethyl)-2-methoxybenzenesulphonamide
Formula: C₉H₁₂BrNO₄S
MDL Number: MFCD08056375
CAS Number: 871269-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR