2-Bromo-N-(2,4-difluorophenyl)benzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CS-9436-5G CS-9436-1G
COBBCS-9436-5GEA 0 GBP
COBBCS-9436-5G COBBCS-9436-1G
2-Bromo-N-(2,4-difluorophenyl)benzamide
2-Bromo-N-(2,4-difluorophenyl)benzamide
Formula: C₁₃H₈BrF₂NO
MDL Number: MFCD00751577
CAS Number: 314025-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR