4-Bromo-2-fluoro-N-methylbenzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4513-5G CA-4513-25G CA-4513-1G
COBBCA-4513-5GEA 0 GBP
COBBCA-4513-5G COBBCA-4513-25G COBBCA-4513-1G
4-Bromo-2-fluoro-N-methylbenzamide
4-Bromo-2-fluoro-N-methylbenzamide
Formula: C₈H₇BrFNO
MW: 232.0497 g/mol
MDL Number: MFCD09878362
CAS Number: 749927-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR