3-Bromo-N-butyl-4-fluorobenzamide
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4615-5G
CA-4615-25G
CA-4615-1G
COBBCA-4615-5GEA
0
GBP
COBBCA-4615-5G
COBBCA-4615-25G
COBBCA-4615-1G
3-Bromo-N-butyl-4-fluorobenzamide
3-Bromo-N-butyl-4-fluorobenzamide
Formula:
C₁₁H₁₃BrFNO |
MDL Number:
MFCD11053800 CAS Number: 1065074-05-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...