3-Bromo-N-tert-butyl-4-fluorobenzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4508-5G CA-4508-25G CA-4508-1G
COBBCA-4508-5GEA 0 GBP
COBBCA-4508-5G COBBCA-4508-25G COBBCA-4508-1G
3-Bromo-N-tert-butyl-4-fluorobenzamide
3-Bromo-N-tert-butyl-4-fluorobenzamide
Formula: C₁₁H₁₃BrFNO
MDL Number: MFCD10699644
CAS Number: 1033201-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR